3,6-Dibromobenzene-1,2-diol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3,6-Dibromo-1,2-benzenediol

207424-1G 207424-500MG 207424-5G 207424-25G
77466-702EA 385.22 USD
77466-702 77479-434 77489-708 77500-010
3,6-Dibromobenzene-1,2-diol ≥95%
3,6-Dibromobenzene-1,2-diol

Precaución: For research use only

Formula: C₆H₄Br₂O₂
MW: 267.90 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28385619
CAS Number: 123433-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR