2-Chloro-5-fluoro-4-(trifluoromethyl)benzenamine ≥95%

Supplier: AOBChem USA

Warning

206864-1G 206864-500MG 206864-5G 206864-25G
77466-698EA 196.73 USD
77466-698 77479-430 77489-704 77500-006
2-Chloro-5-fluoro-4-(trifluoromethyl)benzenamine ≥95%
2-Chloro-5-fluoro-4-(trifluoromethyl)benzenamine

Precaución: For research use only

Formula: C₇H₄ClF₄N
MW: 213.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28752558
CAS Number: 1805524-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR