1-(2,3-Dihydrothieno[3,4-b]-1,4-dioxin-5-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

205730-1G 205730-500MG 205730-5G
77466-694EA 522.21 USD
77466-694 77479-426 77489-700
1-(2,3-Dihydrothieno[3,4-b]-1,4-dioxin-5-yl)-2,2-difluoroethanone ≥95%
1-(2,3-Dihydrothieno[3,4-b]-1,4-dioxin-5-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₆F₂O₃S
MW: 220.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34866139
CAS Number: 2025709-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR