2-Chloro-5-methoxy-4-methylpyridine ≥95%

Supplier: AOBChem USA

Warning

20555-1G 20555-500MG 20555-5G 20555-25G
77466-690EA 258.53 USD
77466-690 77479-422 77489-696 77500-002
2-Chloro-5-methoxy-4-methylpyridine ≥95%
2-Chloro-5-methoxy-4-methylpyridine

Precaución: For research use only

Formula: C₇H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD16610593
CAS Number: 1227593-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR