7-Methyl-1H-indazole-5-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

205217-1G 205217-500MG 205217-5G 205217-25G
77466-686EA 129.78 USD
77466-686 77479-418 77489-690 77499-998
7-Methyl-1H-indazole-5-carboxylic acid ≥97%
7-Methyl-1H-indazole-5-carboxylic acid

Precaución: For research use only

Formula: C₉H₈N₂O₂
MW: 176.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14585353
CAS Number: 1031417-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR