6-Hydroxy-4H-chromen-4-one ≥95%

Supplier: AOBChem USA

Warning

205200-1G 205200-500MG 205200-5G
77466-682EA 775.59 USD
77466-682 77479-414 77489-686
6-Hydroxy-4H-chromen-4-one ≥95%
6-Hydroxy-4H-chromen-4-one

Precaución: For research use only

Formula: C₉H₆O₃
MW: 162.1421 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07808000
CAS Number: 38445-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR