2-Fluoro-5-hydroxypyridine-4-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

20466-1G 20466-500MG 20466-5G 20466-25G
77466-678EA 284.28 USD
77466-678 77479-410 77489-682 77499-994
2-Fluoro-5-hydroxypyridine-4-carboxylic acid ≥95%
2-Fluoro-5-hydroxypyridine-4-carboxylic acid

Precaución: For research use only

Formula: C₆H₄FNO₃
MW: 157.099 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611053
CAS Number: 1211515-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR