3-Pyridinecarboxaldehyde, 6-chloro-2-(trifluoromethyl)- ≥95%

Supplier: AOBChem USA

Warning

204613-1G 204613-500MG 204613-5G
77466-674EA 535.6 USD
77466-674 77479-408 77489-678
3-Pyridinecarboxaldehyde, 6-chloro-2-(trifluoromethyl)- ≥95%
3-Pyridinecarboxaldehyde, 6-chloro-2-(trifluoromethyl)-

Precaución: For research use only

Formula: C₇H₃ClF₃NO
MW: 209.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16610713
CAS Number: 1227581-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR