2,3-Dihydro-1-benzofuran-5-ol ≥97%

Supplier: AOBChem USA

Warning

20391-1G 20391-500MG 20391-5G 20391-25G
77466-670EA 161.71 USD
77466-670 77479-404 77489-674 77499-988
2,3-Dihydro-1-benzofuran-5-ol ≥97%
2,3-Dihydro-1-benzofuran-5-ol

Precaución: For research use only

Formula: C₈H₈O₂
Storage Temperature: Ambient
MDL Number: MFCD09029838
CAS Number: 40492-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR