3-(4-Fluorophenyl)-2-propynoic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-(4-Fluorophenyl)propiolic acid

203519-1G 203519-500MG 203519-5G 203519-25G
77466-666EA 125.66 USD
77466-666 77479-400 77489-670 77499-984
3-(4-Fluorophenyl)-2-propynoic acid ≥95%
3-(4-Fluorophenyl)-2-propynoic acid

Precaución: For research use only

Formula: C₉H₅FO₂
MW: 164.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11934514
CAS Number: 706-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR