1-Bromo-3-(methoxymethoxy)-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

20339-1G 20339-500MG 20339-5G 20339-25G
77466-662EA 82.4 USD
77466-662 77479-396 77489-666 77499-980
1-Bromo-3-(methoxymethoxy)-5-methylbenzene ≥95%
1-Bromo-3-(methoxymethoxy)-5-methylbenzene

Precaución: For research use only

Formula: C₉H₁₁BrO₂
MW: 231.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631783
CAS Number: 2379322-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR