1-[2-(3-Methylphenyl)ethynyl]cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

203240-1G 203240-500MG 203240-5G
77466-658EA 576.8 USD
77466-658 77479-392 77489-662
1-[2-(3-Methylphenyl)ethynyl]cyclopentanol ≥95%
1-[2-(3-Methylphenyl)ethynyl]cyclopentanol

Precaución: For research use only

Formula: C₁₄H₁₆O
MW: 200.28 g/mol
Storage Temperature: Ambient
CAS Number: 1417506-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR