2-(3,6-Dichloro-2-fluorophenyl)butan-2-ol ≥95%

Supplier: AOBChem USA

Warning

203035-1G 203035-500MG 203035-5G 203035-25G
77466-654EA 415.09 USD
77466-654 77479-388 77489-654 77499-974
2-(3,6-Dichloro-2-fluorophenyl)butan-2-ol ≥95%
2-(3,6-Dichloro-2-fluorophenyl)butan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₁Cl₂FO
MW: 237.098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34865698
CAS Number: 2969433-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR