2-(5-Chloro-2-fluoro-3-methylphenyl)butan-2-ol ≥95%

Supplier: AOBChem USA

Warning

202952-1G 202952-500MG 202952-5G 202952-25G
77466-650EA 415.09 USD
77466-650 77479-384 77489-650 77499-970
2-(5-Chloro-2-fluoro-3-methylphenyl)butan-2-ol ≥95%
2-(5-Chloro-2-fluoro-3-methylphenyl)butan-2-ol

Precaución: For research use only

Formula: C₁₁H₁₄ClFO
MW: 216.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34865615
CAS Number: 2969351-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR