(6-Bromo-2-fluoro-3-(methylthio)phenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

202934-1G 202934-500MG 202934-5G 202934-25G
77466-646EA 415.09 USD
77466-646 77479-380 77489-646 77499-966
(6-Bromo-2-fluoro-3-(methylthio)phenyl)(cyclohexyl)methanol ≥95%
(6-Bromo-2-fluoro-3-(methylthio)phenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₈BrFOS
MW: 333.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34865597
CAS Number: 2969467-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR