(6-Bromo-2-fluoro-3-propoxyphenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

202925-1G 202925-500MG 202925-5G 202925-25G
77466-642EA 415.09 USD
77466-642 77479-376 77489-642 77499-962
(6-Bromo-2-fluoro-3-propoxyphenyl)(cyclohexyl)methanol ≥95%
(6-Bromo-2-fluoro-3-propoxyphenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₆H₂₂BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34865588
CAS Number: 2969423-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR