(2-Bromo-6-fluoro-3-propoxyphenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

202923-1G 202923-500MG 202923-5G 202923-25G
77466-638EA 415.09 USD
77466-638 77479-372 77489-638 77499-958
(2-Bromo-6-fluoro-3-propoxyphenyl)(cyclohexyl)methanol ≥95%
(2-Bromo-6-fluoro-3-propoxyphenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₆H₂₂BrFO₂
MW: 345.247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34865586
CAS Number: 2969407-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR