Cyclohexyl(3,6-dibromo-2-fluorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

202919-1G 202919-500MG 202919-5G 202919-25G
77466-634EA 415.09 USD
77466-634 77479-368 77489-634 77499-954
Cyclohexyl(3,6-dibromo-2-fluorophenyl)methanol ≥95%
Cyclohexyl(3,6-dibromo-2-fluorophenyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₅Br₂FO
MW: 366.064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34865582
CAS Number: 2969380-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR