(6-Bromo-3-chloro-2-fluorophenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

202915-1G 202915-500MG 202915-5G 202915-25G
77466-630EA 415.09 USD
77466-630 77479-364 77489-630 77499-950
(6-Bromo-3-chloro-2-fluorophenyl)(cyclohexyl)methanol ≥95%
(6-Bromo-3-chloro-2-fluorophenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₅BrClFO
MW: 321.613 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34865578
CAS Number: 2969431-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR