Cyclohexyl(2-fluoro-5-iodophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

202878-1G 202878-500MG 202878-5G 202878-25G
77466-622EA 415.09 USD
77466-622 77479-356 77489-622 77499-942
Cyclohexyl(2-fluoro-5-iodophenyl)methanol ≥95%
Cyclohexyl(2-fluoro-5-iodophenyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₆FIO
MW: 334.168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34865541
CAS Number: 2969423-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR