(5-Chloro-2-fluoro-3-methylphenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

202861-1G 202861-500MG 202861-5G 202861-25G
77466-618EA 415.09 USD
77466-618 77479-352 77489-618 77499-938
(5-Chloro-2-fluoro-3-methylphenyl)(cyclohexyl)methanol ≥95%
(5-Chloro-2-fluoro-3-methylphenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₈ClFO
MW: 256.744 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34865524
CAS Number: 2969407-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR