6-Bromo-1-chloro-2-methoxynaphthalene ≥97%

Supplier: AOBChem USA

Warning

20215-1G 20215-500MG 20215-5G 20215-25G
77466-610EA 311.06 USD
77466-610 77479-344 77489-610 77499-932
6-Bromo-1-chloro-2-methoxynaphthalene ≥97%
6-Bromo-1-chloro-2-methoxynaphthalene

Precaución: For research use only

Formula: C₁₁H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD22543652
CAS Number: 92455-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR