4-Bromo-2,6-difluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

20200-1G 20200-500MG 20200-5G 20200-25G
77466-606EA 120.51 USD
77466-606 77479-340 77489-606 77499-928
4-Bromo-2,6-difluorobenzamide ≥97%
4-Bromo-2,6-difluorobenzamide

Precaución: For research use only

Formula: C₇H₄BrF₂NO
Storage Temperature: Ambient
MDL Number: MFCD13689151
CAS Number: 840481-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR