4-Chloro-1-methyl-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

20181-1G 20181-5G 20181-25G
77466-598EA 60.77 USD
77466-598 77489-596 77499-920
4-Chloro-1-methyl-1H-indazole ≥95%
4-Chloro-1-methyl-1H-indazole

Precaución: For research use only

Formula: C₈H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD09998749
CAS Number: 162502-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR