(3-Bromo-5-trifluoromethylpyridin-2-yl)dimethylamine ≥95%

Supplier: AOBChem USA

Warning

20113-1G 20113-500MG 20113-5G 20113-25G
77466-594EA 101.97 USD
77466-594 77479-332 77489-590 77499-914
(3-Bromo-5-trifluoromethylpyridin-2-yl)dimethylamine ≥95%
(3-Bromo-5-trifluoromethylpyridin-2-yl)dimethylamine

Precaución: For research use only

Formula: C₈H₈BrF₃N₂
Storage Temperature: Ambient
MDL Number: MFCD18384845
CAS Number: 216765-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR