4-Bromo-3-fluoro-5-(trifluoromethyl)aniline ≥95%

Supplier: AOBChem USA

Warning

200390-1G 200390-500MG 200390-5G 200390-25G
77466-586EA 328.57 USD
77466-586 77479-328 77489-580 77499-904
4-Bromo-3-fluoro-5-(trifluoromethyl)aniline ≥95%
4-Bromo-3-fluoro-5-(trifluoromethyl)aniline

Precaución: For research use only

Formula: C₇H₄BrF₄N
MW: 258.0109 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28348545
CAS Number: 1533426-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR