1-Bromo-3,4-difluoro-2-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

19971-1G 19971-500MG 19971-5G 19971-25G
77466-582EA 253.38 USD
77466-582 77479-324 77489-576 77499-900
1-Bromo-3,4-difluoro-2-isopropoxybenzene ≥97%
1-Bromo-3,4-difluoro-2-isopropoxybenzene

Precaución: For research use only

Formula: C₉H₉BrF₂O
MW: 251.068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17011986
CAS Number: 1242070-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR