6-Methoxy-4-(trifluoromethyl)nicotinaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 6-Methoxy-4-(trifluoromethyl)-3-pyridinecarboxaldehyde

199678-1G 199678-500MG 199678-5G 199678-25G
77466-578EA 268.83 USD
77466-578 77479-320 77489-572 77499-898
6-Methoxy-4-(trifluoromethyl)nicotinaldehyde ≥95%
6-Methoxy-4-(trifluoromethyl)nicotinaldehyde

Precaución: For research use only

Formula: C₈H₆F₃NO₂
MW: 205.134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18258322
CAS Number: 1005171-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR