2-Bromo-5-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

19869-1G 19869-500MG 19869-5G 19869-25G
77466-570EA 204.97 USD
77466-570 77479-312 77489-564 77499-890
2-Bromo-5-(trifluoromethyl)benzamide ≥97%
2-Bromo-5-(trifluoromethyl)benzamide

Precaución: For research use only

Formula: C₈H₅BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD09999463
CAS Number: 1369843-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR