2-Chloro-6-(trifluoromethyl)anisole ≥95%

Supplier: AOBChem USA

Warning

19799-1G 19799-500MG 19799-5G 19799-25G
77466-566EA 426.42 USD
77466-566 77479-308 77489-558 77499-884
2-Chloro-6-(trifluoromethyl)anisole ≥95%
2-Chloro-6-(trifluoromethyl)anisole

Precaución: For research use only

Formula: C₈H₆ClF₃O
MW: 210.581 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185463
CAS Number: 1214342-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR