3-(4-Methoxy-3-methylphenyl)-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

197875-1G 197875-500MG 197875-5G 197875-25G
77466-562EA 392.43 USD
77466-562 77479-304 77489-552 77499-876
3-(4-Methoxy-3-methylphenyl)-1H-pyrazole ≥97%
3-(4-Methoxy-3-methylphenyl)-1H-pyrazole

Precaución: For research use only

Formula: C₁₁H₁₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD34864242
CAS Number: 1586700-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR