3-Ethyl-2-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

197782-1G 197782-500MG 197782-5G
77466-558EA 810.61 USD
77466-558 77479-300 77489-548
3-Ethyl-2-methylbenzoic acid ≥95%
3-Ethyl-2-methylbenzoic acid

Precaución: For research use only

Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD20650625
CAS Number: 55262-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR