4-Benzoylbenzaldehyde ≥95%
Supplier: AOBChem USA
|
Warning
|
196998-1G
196998-500MG
196998-5G
196998-25G
77466-554EA
229.69
USD
77466-554
77479-296
77489-542
77499-870
4-Benzoylbenzaldehyde ≥95%
4-Benzoylbenzaldehyde
Precaución: For research use only
Formula:
C₁₄H₁₀O₂ Storage Temperature: Ambient |
MDL Number:
MFCD02292563 CAS Number: 20912-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...