1,2-Difluoro-3-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

19665-1G 19665-500MG 19665-5G 19665-25G
77466-550EA 176.13 USD
77466-550 77479-292 77489-538 77499-866
1,2-Difluoro-3-(methoxymethoxy)benzene ≥97%
1,2-Difluoro-3-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD30179093
CAS Number: 749230-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR