4-[(4-Methoxyphenyl)methoxy]-2-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

196525-1G 196525-500MG 196525-5G 196525-25G
77466-546EA 415.09 USD
77466-546 77479-288 77489-534 77499-862
4-[(4-Methoxyphenyl)methoxy]-2-methylbenzoic acid ≥95%
4-[(4-Methoxyphenyl)methoxy]-2-methylbenzoic acid

Precaución: For research use only

Formula: C₁₆H₁₆O₄
MW: 272.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34863853
CAS Number: 1537620-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR