4-[(4-Methoxyphenyl)methoxy]-2-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

196427-1G 196427-500MG 196427-5G 196427-25G
77466-542EA 415.09 USD
77466-542 77479-284 77489-530 77499-858
4-[(4-Methoxyphenyl)methoxy]-2-methylbenzaldehyde ≥95%
4-[(4-Methoxyphenyl)methoxy]-2-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₆H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD22840708
CAS Number: 1514811-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR