3-Bromo-2,6-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

195918-1G 195918-500MG 195918-5G 195918-25G
77466-538EA 288.4 USD
77466-538 77479-280 77489-526 77499-856
3-Bromo-2,6-dimethylbenzaldehyde ≥97%
3-Bromo-2,6-dimethylbenzaldehyde

Precaución: For research use only

Formula: C₉H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD28637585
CAS Number: 106965-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR