4-Bromo-2-chloro-1-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

19550-1G 19550-500MG 19550-5G 19550-25G
77466-530EA 219.39 USD
77466-530 77479-272 77489-518 77499-852
4-Bromo-2-chloro-1-(methoxymethoxy)benzene ≥95%
4-Bromo-2-chloro-1-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₈BrClO₂
MW: 251.505 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29946732
CAS Number: 1301146-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR