1-[4-Bromo-2-fluoro-3-(trifluoromethyl)phenyl]ethanone ≥95%

Supplier: AOBChem USA

Warning

195248-1G 195248-500MG 195248-5G
77466-526EA 1240.12 USD
77466-526 77479-268 77489-514
1-[4-Bromo-2-fluoro-3-(trifluoromethyl)phenyl]ethanone ≥95%
1-[4-Bromo-2-fluoro-3-(trifluoromethyl)phenyl]ethanone

Precaución: For research use only

Formula: C₉H₅BrF₄O
MW: 285.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34181532
CAS Number: 2384288-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR