2,3-Difluoro-1-(methylthio)-4-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

194741-1G 194741-500MG 194741-5G 194741-25G
77466-522EA 415.09 USD
77466-522 77479-264 77489-510 77499-848
2,3-Difluoro-1-(methylthio)-4-(trifluoromethyl)benzene ≥95%
2,3-Difluoro-1-(methylthio)-4-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₈H₅F₅S
MW: 228.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34863115
CAS Number: 2705649-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR