2,3-Difluoro-4-iodobenzotrifluoride ≥95%

Supplier: AOBChem USA

Warning

194652-1G 194652-500MG 194652-5G
77466-518EA 488.22 USD
77466-518 77479-260 77489-506
2,3-Difluoro-4-iodobenzotrifluoride ≥95%
2,3-Difluoro-4-iodobenzotrifluoride

Precaución: For research use only

Formula: C₇H₂F₅I
Storage Temperature: Ambient
MDL Number: MFCD31568564
CAS Number: 2227272-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR