4-Bromo-α-(1,1-dimethylethyl)-5-methyl-2-thiophenemethanol ≥95%

Supplier: AOBChem USA

Warning

193819-1G 193819-500MG 193819-5G
77466-510EA 515 USD
77466-510 77479-252 77489-498
4-Bromo-α-(1,1-dimethylethyl)-5-methyl-2-thiophenemethanol ≥95%
4-Bromo-α-(1,1-dimethylethyl)-5-methyl-2-thiophenemethanol

Precaución: For research use only

Formula: C₁₀H₁₅BrOS
MW: 263.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34862806
CAS Number: 1600794-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR