2-Bromo-1-fluoro-4-propoxybenzene ≥95%

Supplier: AOBChem USA

Warning

192636-1G 192636-500MG 192636-5G 192636-25G
77466-498EA 328.57 USD
77466-498 77479-242 77489-482 77499-830
2-Bromo-1-fluoro-4-propoxybenzene ≥95%
2-Bromo-1-fluoro-4-propoxybenzene

Precaución: For research use only

Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26024419
CAS Number: 1547625-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR