3-Bromo-4-isopropylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

192512-1G 192512-500MG 192512-5G
77466-494EA 740.57 USD
77466-494 77479-238 77489-478
3-Bromo-4-isopropylbenzaldehyde ≥97%
3-Bromo-4-isopropylbenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD28606919
CAS Number: 461425-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR