4-(methylsulfonyl)thiomorpholine ≥97%

Supplier: AOBChem USA

Warning

192457-1G 192457-500MG 192457-5G 192457-25G
77466-490EA 397.58 USD
77466-490 77479-234 77489-472 77499-822
4-(methylsulfonyl)thiomorpholine ≥97%
4-(methylsulfonyl)thiomorpholine

Precaución: For research use only

Formula: C₅H₁₁NO₂S₂
MW: 181.2763 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20358270
CAS Number: 98137-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR