1-Propanone, 1-(5-methyl-3-pyridinyl)- ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
1-(5-Methylpyridin-3-yl)propan-1-one
192310-1G
192310-500MG
192310-5G
192310-25G
77466-486EA
202.91
USD
77466-486
77479-230
77489-468
77499-816
1-Propanone, 1-(5-methyl-3-pyridinyl)- ≥95%
1-Propanone, 1-(5-methyl-3-pyridinyl)-
Precaución: For research use only
Formula:
C₉H₁₁NO MW: 149.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24191863 CAS Number: 1174985-74-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...