Ethanone, 1-(6-methyl-1,3-benzodioxol-5-yl)- ≥95%

Supplier: AOBChem USA

Warning

191712-1G 191712-500MG 191712-5G
77466-482EA 757.05 USD
77466-482 77479-226 77489-462
Ethanone, 1-(6-methyl-1,3-benzodioxol-5-yl)- ≥95%
Ethanone, 1-(6-methyl-1,3-benzodioxol-5-yl)-

Precaución: For research use only

Formula: C₁₀H₁₀O₃
MW: 178.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00225575
CAS Number: 96543-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR