5-Bromo-2-ethyl-benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

19037-1G 19037-500MG 19037-5G 19037-25G
77466-474EA 104.03 USD
77466-474 77479-218 77489-456 77499-804
5-Bromo-2-ethyl-benzaldehyde ≥95%
5-Bromo-2-ethyl-benzaldehyde

Precaución: For research use only

Formula: C₉H₉BrO
MW: 213.071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16619424
CAS Number: 946001-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR