2-[(2-Bromophenyl)methyl]oxirane ≥95%

Supplier: AOBChem USA

Warning

190323-1G 190323-500MG 190323-5G
77466-470EA 574.74 USD
77466-470 77479-214 77489-452
2-[(2-Bromophenyl)methyl]oxirane ≥95%
2-[(2-Bromophenyl)methyl]oxirane

Precaución: For research use only

Formula: C₉H₉BrO
MW: 213.07 g/mol
Storage Temperature: Ambient
CAS Number: 62826-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR