4-Cyano-α-hydroxybenzeneacetic acid ≥97%

Supplier: AOBChem USA

Warning

189692-1G 189692-500MG 189692-5G
77466-462EA 592.25 USD
77466-462 77479-206 77489-444
4-Cyano-α-hydroxybenzeneacetic acid ≥97%
4-Cyano-α-hydroxybenzeneacetic acid

Precaución: For research use only

Formula: C₉H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD20658797
CAS Number: 631897-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR